Assay Details
| Gene Symbol: AT4G13360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117410.7 | NP_193072.2 | 9 - 10 | 1275 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G13360| Gene Symbol: AT4G13360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117410.7 | NP_193072.2 | 9 - 10 | 1275 | 80 | |
Set Membership: |
|