Assay Details
| Gene Symbol: IIL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117417.4 | NP_567405.1 | 5 - 6 | 768 | 66 | |
| GenBank mRNA | AY045842.1 | - | 5 - 6 | 642 | 66 | |
| AY117151.1 | - | 5 - 6 | 573 | 66 | ||
More Information
Set Membership: |
|

IIL1
Genomic Map