Assay Details
| Gene Symbol: SUVH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036548.2 | NP_001031625.1 | 1 | 2269 | 68 | |
| NM_117420.3 | NP_193082.1 | 2 | 2150 | 68 | ||
| GenBank mRNA | AF344452.1 | - | 1 | 1870 | 68 | |
| AK317297.1 | - | 1 | 2271 | 68 | ||
More Information
Set Membership: |
|

SUVH9
Genomic Map