Assay Details
| Gene Symbol: SMAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117426.4 | NP_567406.1 | 1 | 365 | 142 | |
| GenBank mRNA | AF361862.1 | - | 1 | 218 | 142 | |
| AY088143.1 | - | 1 | 242 | 142 | ||
More Information
Set Membership: |
|

SMAP1
Genomic Map