Assay Details
| Gene Symbol: NOV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117449.4 | NP_193111.2 | 12 - 13 | 7297 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOV| Gene Symbol: NOV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117449.4 | NP_193111.2 | 12 - 13 | 7297 | 84 | |
Set Membership: |
|