Assay Details
| Gene Symbol: WRNEXO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117461.2 | NP_193123.2 | 2 - 3 | 467 | 83 | |
| NM_202814.2 | NP_974543.1 | 2 - 3 | 482 | 83 | ||
| GenBank mRNA | AF531179.1 | - | 2 - 3 | 472 | 83 | |
| AJ404476.1 | - | 2 - 3 | 467 | 83 | ||
| BT010908.1 | - | 2 - 3 | 432 | 83 | ||
More Information
Set Membership: |
|

WRNEXO
Genomic Map