Assay Details
| Gene Symbol: SBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203794.2 | NP_001190723.1 | 5 - 6 | 853 | 111 | |
| NM_117478.6 | NP_193139.1 | 4 - 5 | 986 | 111 | ||
| GenBank mRNA | AY062634.1 | - | 4 - 5 | 859 | 111 | |
| AY128727.1 | - | 4 - 5 | 811 | 111 | ||
More Information
Set Membership: |
|

SBP1
Genomic Map