Assay Details
| Gene Symbol: PDS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340907.1 | NP_001319934.1 | 10 - 11 | 1502 | 146 | |
| NM_001340908.1 | NP_001329033.1 | 9 - 10 | 1350 | 146 | ||
| NM_117498.4 | NP_193157.1 | 10 - 11 | 1544 | 146 | ||
| GenBank mRNA | AF360196.1 | - | 10 - 11 | 1322 | 146 | |
| AY040007.1 | - | 9 - 10 | 1191 | 146 | ||
| L16237.1 | - | 10 - 11 | 1318 | 146 | ||
More Information
Set Membership: |
|

PDS3
Genomic Map