Assay Details
| Gene Symbol: PDS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340907.1 | NP_001319934.1 | 2 - 3 | 479 | 72 | |
| NM_001340908.1 | NP_001329033.1 | 1 - 2 | 327 | 72 | ||
| NM_117498.4 | NP_193157.1 | 2 - 3 | 521 | 72 | ||
| GenBank mRNA | AF360196.1 | - | 2 - 3 | 299 | 72 | |
| AY040007.1 | - | 1 - 2 | 168 | 72 | ||
| L16237.1 | - | 2 - 3 | 295 | 72 | ||
| Gene Symbol: ACT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230311.1 | - | 297 | 72 | ||
More Information
Set Membership: |
|

PDS3
Genomic Map