Assay Details
| Gene Symbol: PDS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340907.1 | NP_001319934.1 | 6 - 7 | 909 | 101 | |
| NM_001340908.1 | NP_001329033.1 | 5 - 6 | 757 | 101 | ||
| NM_117498.4 | NP_193157.1 | 6 - 7 | 951 | 101 | ||
| GenBank mRNA | AF360196.1 | - | 6 - 7 | 729 | 101 | |
| AY040007.1 | - | 5 - 6 | 598 | 101 | ||
| L16237.1 | - | 6 - 7 | 725 | 101 | ||
More Information
Set Membership: |
|

PDS3
Genomic Map