Assay Details
| Gene Symbol: AT4G14230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117500.6 | NP_193159.3 | 7 - 8 | 1516 | 73 | |
| GenBank mRNA | AK226439.1 | - | 7 - 8 | 1451 | 73 | |
| BT023429.1 | - | 7 - 8 | 1450 | 73 | ||
More Information
Set Membership: |
|

AT4G14230
Genomic Map