Assay Details
| Gene Symbol: AT4G14280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117505.4 | NP_193164.2 | 1 - 2 | 230 | 68 | |
| GenBank mRNA | DQ446835.1 | - | 1 - 2 | 179 | 68 | |
More Information
Set Membership: |
|

AT4G14280
Genomic Map