Assay Details
| Gene Symbol: AT4G14280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117505.4 | NP_193164.2 | 2 - 3 | 2020 | 111 | |
| GenBank mRNA | DQ446835.1 | - | 2 - 3 | 1969 | 111 | |
More Information
Set Membership: |
|

AT4G14280
Genomic Map