Assay Details
| Gene Symbol: CKI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340918.1 | NP_001328596.1 | 11 - 12 | 1262 | 57 | |
| NM_117511.5 | NP_193170.1 | 11 - 12 | 1262 | 57 | ||
| GenBank mRNA | AF250343.1 | - | 11 - 12 | 1086 | 57 | |
| AY052364.1 | - | 11 - 12 | 1232 | 57 | ||
| AY060528.1 | - | 11 - 12 | 932 | 57 | ||
| AY943850.1 | - | 11 - 12 | 932 | 57 | ||
| X78818.1 | - | 1103 | 57 | |||
More Information
Set Membership: |
|

CKI1
Genomic Map