Assay Details
| Gene Symbol: CFM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117531.4 | NP_193187.3 | 8 - 9 | 2623 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFM3B| Gene Symbol: CFM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117531.4 | NP_193187.3 | 8 - 9 | 2623 | 88 | |
Set Membership: |
|