Assay Details
| Gene Symbol: CIPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117538.2 | NP_193194.1 | 1 | 1008 | 82 | |
| GenBank mRNA | AY007221.1 | - | 1 | 863 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CIPK4| Gene Symbol: CIPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117538.2 | NP_193194.1 | 1 | 1008 | 82 | |
| GenBank mRNA | AY007221.1 | - | 1 | 863 | 82 | |
Set Membership: |
|