Assay Details
| Gene Symbol: CAM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340951.1 | NP_001329799.1 | 1 - 2 | 164 | 115 | |
| NM_117546.4 | NP_193200.1 | 1 - 2 | 164 | 115 | ||
| GenBank mRNA | AF178074.1 | - | 1 - 2 | 107 | 115 | |
| BT024688.1 | - | 1 - 2 | 76 | 115 | ||
More Information
Set Membership: |
|

CAM8
Genomic Map