Assay Details
| Gene Symbol: TCX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340970.1 | NP_001328548.1 | 5 - 6 | 1524 | 90 | |
| NM_001340971.1 | NP_001328549.1 | 4 - 5 | 1498 | 90 | ||
| NM_117562.7 | NP_193213.5 | 5 - 6 | 1490 | 90 | ||
More Information
Set Membership: |
|

TCX2
Genomic Map