Assay Details
| Gene Symbol: TCX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340970.1 | NP_001328548.1 | 8 - 9 | 1829 | 119 | |
| NM_001340971.1 | NP_001328549.1 | 7 - 8 | 1803 | 119 | ||
| NM_117562.7 | NP_193213.5 | 8 - 9 | 1795 | 119 | ||
More Information
Set Membership: |
|

TCX2
Genomic Map