Assay Details
| Gene Symbol: FH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203807.2 | NP_001190736.2 | 2 - 3 | 896 | 67 | |
| NM_001340996.1 | NP_001328296.1 | 2 - 3 | 896 | 67 | ||
| NM_001340997.1 | NP_001328295.1 | 2 - 3 | 662 | 67 | ||
| NM_117608.5 | NP_193255.6 | 2 - 3 | 605 | 67 | ||
More Information
Set Membership: |
|

FH3
Genomic Map