Assay Details
| Gene Symbol: FH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203807.2 | NP_001190736.2 | 4 - 5 | 1570 | 89 | |
| NM_001340997.1 | NP_001328295.1 | 4 - 5 | 1336 | 89 | ||
| NM_117608.5 | NP_193255.6 | 4 - 5 | 1279 | 89 | ||
More Information
Set Membership: |
|

FH3
Genomic Map