Assay Details
| Gene Symbol: BAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340998.1 | NP_001329368.1 | 5 - 6 | 1504 | 72 | |
| NM_001340999.1 | NP_001329367.1 | 5 - 6 | 1677 | 72 | ||
| NM_117609.3 | NP_567460.1 | 5 - 6 | 1279 | 72 | ||
| NM_179058.1 | NP_849389.1 | 5 - 6 | 1223 | 72 | ||
| GenBank mRNA | AF424573.1 | - | 5 - 6 | 1218 | 72 | |
| AK226353.1 | - | 5 - 6 | 1221 | 72 | ||
| AK316869.1 | - | 5 - 6 | 1219 | 72 | ||
| AY142024.1 | - | 5 - 6 | 1110 | 72 | ||
| M73467.1 | - | 1199 | 72 | |||
More Information
Set Membership: |
|

BAM5
Genomic Map