Assay Details
| Gene Symbol: PEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117622.3 | NP_567462.1 | 12 - 13 | 1705 | 88 | |
| GenBank mRNA | AF062513.1 | - | 12 - 13 | 1694 | 88 | |
More Information
Set Membership: |
|

PEN1
Genomic Map