Assay Details
| Gene Symbol: PEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341033.1 | NP_001328610.1 | 9 - 10 | 1488 | 73 | |
| NM_117622.3 | NP_567462.1 | 9 - 10 | 1499 | 73 | ||
| GenBank mRNA | AF062513.1 | - | 9 - 10 | 1488 | 73 | |
More Information
Set Membership: |
|

PEN1
Genomic Map