Assay Details
| Gene Symbol: CLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117647.3 | NP_193291.1 | 3 - 4 | 985 | 66 | |
| GenBank mRNA | BT002340.1 | - | 3 - 4 | 592 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLA1| Gene Symbol: CLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117647.3 | NP_193291.1 | 3 - 4 | 985 | 66 | |
| GenBank mRNA | BT002340.1 | - | 3 - 4 | 592 | 66 | |
Set Membership: |
|