Assay Details
| Gene Symbol: CLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117647.3 | NP_193291.1 | 6 - 7 | 1838 | 98 | |
| GenBank mRNA | BT002340.1 | - | 6 - 7 | 1445 | 98 | |
More Information
Set Membership: |
|

CLA1
Genomic Map