Assay Details
| Gene Symbol: AT4G15610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117652.4 | NP_567472.1 | 1 - 2 | 627 | 65 | |
| GenBank mRNA | AF326884.1 | - | 1 - 2 | 288 | 65 | |
| AK226478.1 | - | 1 - 2 | 291 | 65 | ||
| AY086594.1 | - | 1 - 2 | 288 | 65 | ||
| AY113938.1 | - | 1 - 2 | 228 | 65 | ||
More Information
Set Membership: |
|

AT4G15610
Genomic Map