Assay Details
| Gene Symbol: AT4G15610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341066.1 | NP_001329530.1 | 2 - 3 | 1177 | 64 | |
| NM_117652.4 | NP_567472.1 | 2 - 3 | 759 | 64 | ||
| GenBank mRNA | AF326884.1 | - | 2 - 3 | 420 | 64 | |
| AK226478.1 | - | 2 - 3 | 423 | 64 | ||
| AY086594.1 | - | 2 - 3 | 420 | 64 | ||
| AY113938.1 | - | 2 - 3 | 360 | 64 | ||
More Information
Set Membership: |
|

AT4G15610
Genomic Map