Assay Details
| Gene Symbol: AT4G15830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117675.3 | NP_567477.1 | 2 - 3 | 359 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G15830| Gene Symbol: AT4G15830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117675.3 | NP_567477.1 | 2 - 3 | 359 | 80 | |
Set Membership: |
|