Assay Details
| Gene Symbol: ESD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341077.1 | NP_001328371.1 | 1 - 2 | 971 | 76 | |
| NM_117680.4 | NP_567478.1 | 1 - 2 | 953 | 76 | ||
| GenBank mRNA | AF386934.1 | - | 1 - 2 | 878 | 76 | |
| AY081548.1 | - | 1 - 2 | 754 | 76 | ||
More Information
Set Membership: |
|

ESD4
Genomic Map