Assay Details
| Gene Symbol: ESD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341077.1 | NP_001328371.1 | 3 - 4 | 1136 | 90 | |
| NM_117680.4 | NP_567478.1 | 3 - 4 | 1118 | 90 | ||
| GenBank mRNA | AF386934.1 | - | 3 - 4 | 1043 | 90 | |
| AY081548.1 | - | 3 - 4 | 919 | 90 | ||
More Information
Set Membership: |
|

ESD4
Genomic Map