Assay Details
| Gene Symbol: ESD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117680.4 | NP_567478.1 | 6 - 7 | 1433 | 134 | |
| GenBank mRNA | AF386934.1 | - | 6 - 7 | 1358 | 134 | |
| AY081548.1 | - | 6 - 7 | 1234 | 134 | ||
More Information
Set Membership: |
|

ESD4
Genomic Map