Assay Details
| Gene Symbol: ESD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341077.1 | NP_001328371.1 | 7 - 8 | 1568 | 99 | |
| NM_117680.4 | NP_567478.1 | 7 - 8 | 1531 | 99 | ||
| GenBank mRNA | AF386934.1 | - | 7 - 8 | 1456 | 99 | |
| AY081548.1 | - | 7 - 8 | 1332 | 99 | ||
More Information
Set Membership: |
|

ESD4
Genomic Map