Assay Details
| Gene Symbol: IMPA-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160767.2 | NP_001154239.1 | 4 - 5 | 1017 | 86 | |
| NM_117708.6 | NP_567485.4 | 4 - 5 | 1017 | 86 | ||
More Information
Set Membership: |
|

IMPA-2
Genomic Map