Assay Details
| Gene Symbol: IMPA-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117708.6 | NP_567485.4 | 9 - 10 | 1894 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IMPA-2| Gene Symbol: IMPA-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117708.6 | NP_567485.4 | 9 - 10 | 1894 | 94 | |
Set Membership: |
|