Assay Details
| Gene Symbol: AT4G16190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117715.3 | NP_567489.1 | 2 - 3 | 826 | 64 | |
| GenBank mRNA | AK226366.1 | - | 2 - 3 | 641 | 64 | |
| AY039556.1 | - | 2 - 3 | 638 | 64 | ||
| AY129473.1 | - | 2 - 3 | 594 | 64 | ||
| AY136316.1 | - | 2 - 3 | 638 | 64 | ||
| BT000733.1 | - | 2 - 3 | 631 | 64 | ||
| Gene Symbol: COR47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230360.1 | - | 737 | 64 | ||
More Information
Set Membership: |
|

AT4G16190
Genomic Map