Assay Details
| Gene Symbol: ECHIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117717.5 | NP_193356.2 | 1 - 2 | 571 | 72 | |
| GenBank mRNA | BT012519.1 | - | 1 - 2 | 430 | 72 | |
| BT014955.1 | - | 1 - 2 | 410 | 72 | ||
More Information
Set Membership: |
|

ECHIA
Genomic Map