Assay Details
| Gene Symbol: ADA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117737.5 | NP_567495.1 | 9 - 10 | 1223 | 94 | |
| NM_202831.3 | NP_974560.1 | 9 - 10 | 1211 | 94 | ||
| NM_202832.2 | NP_974561.1 | 9 - 10 | 1149 | 94 | ||
| GenBank mRNA | AF338770.1 | - | 9 - 10 | 841 | 94 | |
| AY050348.1 | - | 9 - 10 | 1003 | 94 | ||
| AY143911.1 | - | 9 - 10 | 841 | 94 | ||
More Information
Set Membership: |
|

ADA2B
Genomic Map