Assay Details
| Gene Symbol: AT4G16490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117747.7 | NP_567501.4 | 1 - 2 | 1915 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G16490| Gene Symbol: AT4G16490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117747.7 | NP_567501.4 | 1 - 2 | 1915 | 59 | |
Set Membership: |
|