Assay Details
| Gene Symbol: AT4G16560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117755.1 | NP_567505.1 | 1 - 2 | 46 | 66 | |
| Gene Symbol: AT4G16550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341114.1 | NP_001319962.1 | 1 - 2 | 111 | 66 | |
More Information
Set Membership: |
|

AT4G16560
Genomic Map