Assay Details
| Gene Symbol: AT4G16630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117764.5 | NP_193396.3 | 2 - 3 | 884 | 76 | |
| GenBank mRNA | AJ010475.1 | - | 2 - 3 | 884 | 76 | |
| AK228353.1 | - | 2 - 3 | 624 | 76 | ||
More Information
Set Membership: |
|

AT4G16630
Genomic Map