Assay Details
| Gene Symbol: HB-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117780.4 | NP_193411.1 | 1 - 2 | 340 | 92 | |
| GenBank mRNA | AF375453.1 | - | 1 - 2 | 249 | 92 | |
| AY081747.1 | - | 1 - 2 | 163 | 92 | ||
| X68145.1 | - | 340 | 92 | |||
More Information
Set Membership: |
|

HB-2
Genomic Map