Assay Details
| Gene Symbol: CLPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117808.3 | NP_567521.1 | 3 - 4 | 601 | 73 | |
| GenBank mRNA | AK117499.1 | - | 3 - 4 | 542 | 73 | |
| AY087492.1 | - | 3 - 4 | 587 | 73 | ||
More Information
Set Membership: |
|

CLPR4
Genomic Map