Assay Details
| Gene Symbol: CLPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117808.3 | NP_567521.1 | 4 - 5 | 734 | 66 | |
| GenBank mRNA | AK117499.1 | - | 4 - 5 | 675 | 66 | |
| AY087492.1 | - | 4 - 5 | 720 | 66 | ||
More Information
Set Membership: |
|

CLPR4
Genomic Map