Assay Details
| Gene Symbol: CT-BMY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117813.3 | NP_567523.1 | 1 - 2 | 682 | 69 | |
| GenBank mRNA | AJ250341.1 | - | 1 - 2 | 577 | 69 | |
| AY087592.1 | - | 1 - 2 | 614 | 69 | ||
More Information
Set Membership: |
|

CT-BMY
Genomic Map