Assay Details
| Gene Symbol: NS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341198.1 | NP_001328216.1 | 3 - 4 | 781 | 82 | |
| NM_117835.5 | NP_193462.1 | 3 - 4 | 613 | 82 | ||
| GenBank mRNA | AY078967.1 | - | 3 - 4 | 590 | 82 | |
More Information
Set Membership: |
|

NS1
Genomic Map