Assay Details
| Gene Symbol: ERF-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117855.6 | NP_567530.4 | 1 | 896 | 78 | |
| GenBank mRNA | AY058174.1 | - | 1 | 774 | 78 | |
| AY062533.1 | - | 1 | 770 | 78 | ||
| BT002578.1 | - | 1 | 694 | 78 | ||
More Information
Set Membership: |
|

ERF-1
Genomic Map