Assay Details
| Gene Symbol: UCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117857.3 | NP_193484.1 | 2 - 3 | 395 | 95 | |
| GenBank mRNA | AK118904.1 | - | 2 - 3 | 334 | 95 | |
| BT005488.1 | - | 2 - 3 | 212 | 95 | ||
More Information
Set Membership: |
|

UCH3
Genomic Map