Assay Details
| Gene Symbol: UCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117857.3 | NP_193484.1 | 7 - 8 | 719 | 72 | |
| GenBank mRNA | AK118904.1 | - | 7 - 8 | 658 | 72 | |
| BT005488.1 | - | 7 - 8 | 536 | 72 | ||
More Information
Set Membership: |
|

UCH3
Genomic Map