Assay Details
| Gene Symbol: RAB1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117859.4 | NP_193486.1 | 7 - 8 | 768 | 60 | |
| GenBank mRNA | AF127134.1 | - | 7 - 8 | 669 | 60 | |
| AK227290.1 | - | 7 - 8 | 679 | 60 | ||
| AY052204.1 | - | 7 - 8 | 677 | 60 | ||
More Information
Set Membership: |
|

RAB1C
Genomic Map